Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-3-oxo-2,3-dihydro-4H-1,4-benzoxazine-4-propionic acid, 97%

$70.36 - $235.05

Chemical Identifiers

Chemical Name or Material 6-Chloro-3-oxo-2,3-dihydro-4H-1,4-benzoxazine-4-propionic acid
CAS 351003-03-7
Molecular Formula C11H10ClNO4
Formula Weight 255.65
MDL Number MFCD03424566
Synonym 6-Chloro-2,3-dihydro-3-oxo-4H-1,4-benzoxazine-4-propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2786303

  SDS
alfa aesar™
H2786303
1g Each for $70.36

AAH2786306

  SDS
alfa aesar™
H2786306
5g Each for $235.05
Description & Specifications

Specifications

Formula Weight 255.65
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Chemical Name or Material 6-Chloro-3-oxo-2,3-dihydro-4H-1,4-benzoxazine-4-propionic acid
Molecular Formula C11H10ClNO4
MDL Number MFCD03424566
Melting Point 146° to 149°C
CAS 351003-03-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Synonym 6-Chloro-2,3-dihydro-3-oxo-4H-1,4-benzoxazine-4-propionic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.