Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-3-oxo-2,3-dihydro-4H-1,4-benzoxazine-4-acetic acid, 96%

$347.53 - $347.53

Chemical Identifiers

Chemical Name or Material 6-Chloro-3-oxo-2,3-dihydro-4H-1,4-benzoxazine-4-acetic acid
CAS 26494-58-6
Molecular Formula C10H8ClNO4
Formula Weight 241.63g/mol
MDL Number MFCD00665821
Synonym (6-Chloro-3-oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)acetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5022203

 
alfa aesar™
H50222-03
Each for $347.53
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 6-Chloro-3-oxo-2,3-dihydro-4H-1,4-benzoxazine-4-acetic acid
CAS 26494-58-6
Assay 96%
Molecular Formula C10H8ClNO4
Formula Weight 241.63g/mol
MDL Number MFCD00665821
Synonym (6-Chloro-3-oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)acetic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.