Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-3-iodo-2-methylpyridine, 95%

$117.84 - $265.19

Chemical Identifiers

Chemical Name or Material 6-Chloro-3-iodo-2-methylpyridine
CAS 249291-79-0
Molecular Formula C6H5ClIN
Formula Weight 253.47
MDL Number MFCD09037461
Synonym 6-Chloro-3-iodo-2-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2765603

  SDS
alfa aesar™
H2765603
1g Each for $265.19

AAH27656MD

  SDS
alfa aesar™
H27656MD
250mg Each for $117.84
Description & Specifications

Specifications

Formula Weight 253.47
MDL Number MFCD09037461
CAS 249291-79-0
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 30° to 33°C
Assay 95%
Molecular Formula C6H5ClIN
Chemical Name or Material 6-Chloro-3-iodo-2-methylpyridine
Synonym 6-Chloro-3-iodo-2-picoline
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.