missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Chloro-3-indolyl-beta-D-glucuronide cyclohexylammonium salt, 98%, Thermo Scientific™

$64.70 - $264.67

Chemical Identifiers

CAS 138182-20-4
Molecular Formula C14H13ClNO7
Molecular Weight (g/mol) 342.71
MDL Number MFCD00153940
InChI Key UFBPRKNLSYGHJJ-BYNIDDHOSA-M
Synonym cyclohexanaminium 2s,3s,4s,5r,6s-6-6-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxytetrahydro-2h-pyran-2-carboxylate, salmon-glca, cyclohexylammonium ion 2s,3s,4s,5r,6s-6-6-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxyoxane-2-carboxylate, c14h14clno7.c6h13n, 6-chloro-3-indolyl b-d-glucuronide cyclohexyl, 6-chloro-3-indolyl beta-d-glucuronide cyclohexylammonium salt, 6-chloro-3-indoxyl-beta-d-glucuronic acid, cyclohexylammonium salt
PubChem CID 16211694
IUPAC Name (2S,3S,4S,5R,6S)-6-[(6-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid;cyclohexanamine
SMILES O[C@@H]1[C@@H](O)[C@H](OC2=CNC3=CC(Cl)=CC=C23)O[C@@H]([C@H]1O)C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB21068MA
View Documents Promotion Details
Thermo Scientific Chemicals
B21068MA
10 mg
Each for $64.70
 
AAB21068MF
View Documents Promotion Details
Thermo Scientific Chemicals
B21068MF
50 mg
Each for $264.67
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

138182-20-4
342.71
UFBPRKNLSYGHJJ-BYNIDDHOSA-M
16211694
O[C@@H]1[C@@H](O)[C@H](OC2=CNC3=CC(Cl)=CC=C23)O[C@@H]([C@H]1O)C([O-])=O
C14H13ClNO7
MFCD00153940
cyclohexanaminium 2s,3s,4s,5r,6s-6-6-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxytetrahydro-2h-pyran-2-carboxylate, salmon-glca, cyclohexylammonium ion 2s,3s,4s,5r,6s-6-6-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxyoxane-2-carboxylate, c14h14clno7.c6h13n, 6-chloro-3-indolyl b-d-glucuronide cyclohexyl, 6-chloro-3-indolyl beta-d-glucuronide cyclohexylammonium salt, 6-chloro-3-indoxyl-beta-d-glucuronic acid, cyclohexylammonium salt
(2S,3S,4S,5R,6S)-6-[(6-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid;cyclohexanamine

Specifications

138182-20-4
MFCD00153940
Light and moisture sensitive
UFBPRKNLSYGHJJ-BYNIDDHOSA-M
(2S,3S,4S,5R,6S)-6-[(6-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid;cyclohexanamine
16211694
98%
C14H13ClNO7
10 mg
cyclohexanaminium 2s,3s,4s,5r,6s-6-6-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxytetrahydro-2h-pyran-2-carboxylate, salmon-glca, cyclohexylammonium ion 2s,3s,4s,5r,6s-6-6-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxyoxane-2-carboxylate, c14h14clno7.c6h13n, 6-chloro-3-indolyl b-d-glucuronide cyclohexyl, 6-chloro-3-indolyl beta-d-glucuronide cyclohexylammonium salt, 6-chloro-3-indoxyl-beta-d-glucuronic acid, cyclohexylammonium salt
O[C@@H]1[C@@H](O)[C@H](OC2=CNC3=CC(Cl)=CC=C23)O[C@@H]([C@H]1O)C([O-])=O
342.71
442.9
6-Chloro-3-indolyl-beta-D-glucuronide cyclohexylammonium salt
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'