Please login to your online account to display your discounted pricing

6-chloro-3-(hydroxymethyl)pyridine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 6-chloro-3-(hydroxymethyl)pyridine
CAS 21543-49-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H6ClNO
Formula Weight 143.57
Synonym 6-chloro-3-pyridinemethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC358090050

 
thermo fisher scientific
AC358090050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC358090010

 
thermo fisher scientific
AC358090010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC358090250

 
thermo fisher scientific
AC358090250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder or Low Melting Solid
Formula Weight 143.57
Quantity 5g
Color White to brown
Density 1.28g/mL
CAS Max % 100.0
Synonym 6-chloro-3-pyridinemethanol
Chemical Name or Material 6-chloro-3-(hydroxymethyl)pyridine
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Molecular Formula C6H6ClNO
CAS 21543-49-7
Assay Percent Range 98%
Infrared Spectrum Authentic
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.