Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-3,4'-bipyridine, 95%

$296.57 - $296.57

Chemical Identifiers

Chemical Name or Material 6-Chloro-3,4'-bipyridine
CAS 79739-22-3
Molecular Formula C10H7ClN2
Formula Weight 190.63g/mol
MDL Number MFCD00137926
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27037ME

 
alfa aesar™
H27037-ME
500mg Each for $296.57
Description & Specifications

Specifications

Quantity 500mg
Quantity 500mg
Chemical Name or Material 6-Chloro-3,4'-bipyridine
CAS 79739-22-3
Assay 95%
Molecular Formula C10H7ClN2
Formula Weight 190.63g/mol
MDL Number MFCD00137926
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.