Please login to your online account to display your discounted pricing

6-Chloro-2H-chromene-3-carbonyl chloride, 95%, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  RDR03724EA

 View more versions of this product

Catalog No. RDR03724EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 229.06
Quantity 10g
Assay 95%
CAS Min % 95.0
Assay Percent Range 95%
Molecular Formula C10H6Cl2O2
Health Hazard 1 GHS Signal Word: Danger
CAS 306935-54-6
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD02091502
Chemical Name or Material 6-Chloro-2H-chromene-3-carbonyl chloride
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

Specifications

Chemical Name or Material 6-Chloro-2H-chromene-3-carbonyl chloride
CAS 306935-54-6
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C10H6Cl2O2
Formula Weight 229.06
MDL Number MFCD02091502
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents