Please login to your online account to display your discounted pricing

6-chloro-2-tetralone, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC427960050

 View more versions of this product

Catalog No. AC427960050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 180.63
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
CAS 17556-18-2
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Molecular Formula C10H9ClO
MDL Number MFCD03788443
Chemical Name or Material 6-chloro-2-tetralone
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.

Specifications

Chemical Name or Material 6-chloro-2-tetralone
CAS 17556-18-2
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H9ClO
Formula Weight 180.63
MDL Number MFCD03788443
Accessories

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.