missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-chloro-2-tetralone, 97%, ACROS Organics™

Chemical Identifiers

CAS 17556-18-2
Molecular Formula C10H9ClO
Molecular Weight (g/mol) 180.631
MDL Number MFCD03788443
InChI Key QQSYTUUAEZUAKL-UHFFFAOYSA-N
Synonym 6-chloro-2-tetralone, 6-chloro-3,4-dihydronaphthalen-2 1h-one, 2 1h-naphthalenone, 6-chloro-3,4-dihydro, 6-chloro-3,4-dihydro-2 1h-naphthalenone, 2 1h-naphthalenone,6-chloro-3,4-dihydro, pubchem17950, 6-chloro-2-tetrarlone, acmc-20a1a6, 6-chloro-beta-tetralone, 6-chloro-3.4-dihydro-2 1h naphthalenone
PubChem CID 4456139
IUPAC Name 6-chloro-3,4-dihydro-1H-naphthalen-2-one
SMILES C1CC2=C(CC1=O)C=CC(=C2)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC427960010 SDS View Documents Acros Organics
AC427960010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC427960050 SDS View Documents Acros Organics
AC427960050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

17556-18-2
180.631
QQSYTUUAEZUAKL-UHFFFAOYSA-N
4456139
C1CC2=C(CC1=O)C=CC(=C2)Cl
C10H9ClO
MFCD03788443
6-chloro-2-tetralone, 6-chloro-3,4-dihydronaphthalen-2 1h-one, 2 1h-naphthalenone, 6-chloro-3,4-dihydro, 6-chloro-3,4-dihydro-2 1h-naphthalenone, 2 1h-naphthalenone,6-chloro-3,4-dihydro, pubchem17950, 6-chloro-2-tetrarlone, acmc-20a1a6, 6-chloro-beta-tetralone, 6-chloro-3.4-dihydro-2 1h naphthalenone
6-chloro-3,4-dihydro-1H-naphthalen-2-one

Specifications

Glass bottle
1g
96.0
97%
MFCD03788443
QQSYTUUAEZUAKL-UHFFFAOYSA-N
6-chloro-3,4-dihydro-1H-naphthalen-2-one
4456139
97%
61.0°C to 65.0°C
17556-18-2
100.0
C10H9ClO
6-chloro-2-tetralone, 6-chloro-3,4-dihydronaphthalen-2 1h-one, 2 1h-naphthalenone, 6-chloro-3,4-dihydro, 6-chloro-3,4-dihydro-2 1h-naphthalenone, 2 1h-naphthalenone,6-chloro-3,4-dihydro, pubchem17950, 6-chloro-2-tetrarlone, acmc-20a1a6, 6-chloro-beta-tetralone, 6-chloro-3.4-dihydro-2 1h naphthalenone
C1CC2=C(CC1=O)C=CC(=C2)Cl
180.631
180.63
6-chloro-2-tetralone
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.