Please login to your online account to display your discounted pricing

Alfa Aesar™ (6-Chloro-2-pyridyl)hydrazine, 95%

$44.61 - $170.30

Chemical Identifiers

Chemical Name or Material (6-Chloro-2-pyridyl)hydrazine
CAS 5193-03-3
Molecular Formula C5H6ClN3
Formula Weight 143.58g/mol
MDL Number MFCD00214724
Synonym 6-Chloropyridin-2-ylhydrazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2724903

 
alfa aesar™
H27249-03
1g Each for $170.30

AAH27249MD

 
alfa aesar™
H27249-MD
250mg Each for $44.61
Description & Specifications

Specifications

Boiling Point 117° to 120°C
Quantity 1g
Chemical Name or Material (6-Chloro-2-pyridyl)hydrazine
CAS 5193-03-3
Assay 95%
Molecular Formula C5H6ClN3
Formula Weight 143.58g/mol
MDL Number MFCD00214724
Synonym 6-Chloropyridin-2-ylhydrazine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.