missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Chloro-2-methyl-2H-1,4-benzoxazin-3(4H)-one 98.0+%, TCI America™
Supplier: TCI America C31385G
Specifications
6-Chloro-2-methyl-2H-1,4-benzoxazin-3(4H)-one | |
174°C | |
MFCD01744501 | |
MGOCBXKZDZRPMK-UHFFFAOYSA-N | |
6-chloro-2-methyl-4H-1,4-benzoxazin-3-one | |
22035 | |
≥98.0% (GC) |
5791-00-4 | |
C9H8ClNO2 | |
5 g | |
CC1C(=O)NC2=C(O1)C=CC(=C2)Cl | |
197.618 | |
197.62 | |
Crystalline Powder |
Chemical Identifiers
5791-00-4 | |
197.618 | |
MGOCBXKZDZRPMK-UHFFFAOYSA-N | |
6-chloro-2-methyl-4H-1,4-benzoxazin-3-one |
C9H8ClNO2 | |
MFCD01744501 | |
22035 | |
CC1C(=O)NC2=C(O1)C=CC(=C2)Cl |
Safety and Handling
RTECSNumber : DM3750000
TSCA : No