Learn More
6-Chloro-2-iodo-3-hydroxypyridine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3312703
Description
2,2'-Dipyridylamine has been obtained by by heating a mixture of 2-aminopyridine and 2-bromo- (chloro-, iodo- ) -pyridine with anhydrous zinc chloride or with barium oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2′-Dipyridylamine has been obtained by by heating a mixture of 2-aminopyridine and 2-bromo- (chloro-, iodo- ) -pyridine with anhydrous zinc chloride or with barium oxide.
Solubility
Slightly Soluble (1.3 g/L) (25°C).
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Specifications
6-Chloro-2-iodo-3-hydroxypyridine | |
191°C to 193°C | |
MFCD07636749 | |
Light sensitive | |
Slightly Soluble (1.3g/L) (25°C). | |
C1=CC(=NC(=C1O)I)Cl | |
255.439 | |
255.44 |
188057-26-3 | |
C5H3ClINO | |
1 g | |
6-chloro-2-iodo-3-hydroxypyridine, 6-chloro-2-iodo-3-pyridinol, 6-chloro-2-iodo-pyridin-3-ol, 3-pyridinol, 6-chloro-2-iodo, 2-iodo-6-chloropyridine-3-ol, 3-pyridinol,6-chloro-2-iodo | |
FWIMPBYPMQSSCD-UHFFFAOYSA-N | |
6-chloro-2-iodopyridin-3-ol | |
23433158 | |
98% |
Chemical Identifiers
188057-26-3 | |
255.439 | |
FWIMPBYPMQSSCD-UHFFFAOYSA-N | |
23433158 | |
C1=CC(=NC(=C1O)I)Cl |
C5H3ClINO | |
MFCD07636749 | |
6-chloro-2-iodo-3-hydroxypyridine, 6-chloro-2-iodo-3-pyridinol, 6-chloro-2-iodo-pyridin-3-ol, 3-pyridinol, 6-chloro-2-iodo, 2-iodo-6-chloropyridine-3-ol, 3-pyridinol,6-chloro-2-iodo | |
6-chloro-2-iodopyridin-3-ol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only