Please login to your online account to display your discounted pricing

6-Chloro-2-hexanone, 98%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  343690050

 View more versions of this product

Catalog No. AC343690050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 134.61
Quantity 5g
Color Clear yellow to brown
EINECS Number 233-546-5
CAS Max % 100.0
Chemical Name or Material 6-Chloro-2-hexanone
Storage Information Product darkens during storage
Molecular Formula C6H11ClO
Packaging Glass bottle
Name Note 98%
Linear Formula H3CCO(CH2)4Cl
MDL Number MFCD00191625
CAS 10226-30-9
CAS Min % 97.5
Infrared Spectrum Authentic
Assay Percent Range 98%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4425 to 1.4445 (20°C, 589nm)

Specifications

Chemical Name or Material 6-Chloro-2-hexanone
Name Note 98%
CAS 10226-30-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H11ClO
Linear Formula H3CCO(CH2)4Cl
Formula Weight 134.61
MDL Number MFCD00191625
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents