Please login to your online account to display your discounted pricing

6-Chloro-2-hexanone, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 6-Chloro-2-hexanone
Name Note 98%
CAS 10226-30-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H11ClO
Linear Formula H3CCO(CH2)4Cl
Formula Weight 134.61
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC343690250

  SDS
thermo fisher scientific
343690250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 134.61
Quantity 25g
Color Clear yellow to brown
EINECS Number 233-546-5
CAS Max % 100.0
Infrared Spectrum Authentic
Linear Formula H3CCO(CH2)4Cl
Refractive Index 1.4425 to 1.4445 (20°C, 589nm)
Name Note 98%
Molecular Formula C6H11ClO
MDL Number MFCD00191625
Chemical Name or Material 6-Chloro-2-hexanone
CAS 10226-30-9
Assay Percent Range 98%
CAS Min % 97.5
Storage Information Product darkens during storage
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.