missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-2-hexanone, 98%

Manufacturer:  Alfa Aesar™ H5346214

 View more versions of this product

Catalog No. AAH5346214



Description

Description

Applications
6-Chloro-2-hexanone in the synthesis of the keto analogs began with 6-chloro-2-hexanone. Substituted 7-(oxoalkyl)theophyllines were synthesized by alkylation of the respective theophyllines with 6-chloro-2-hexanone.

Solubility
Insoluble in water, soluble in chloroform (chloroform, carbon), n-heptane, ethanol, butanol, etc.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

6-Chloro-2-hexanone
86°C (16mmHg)
10226-30-9
MFCD00191625
CMDIDTNMHQUVPE-UHFFFAOYSA-N
6-chlorohexan-2-one
82468
98%
91°C (196°F)
25g
C6H11ClO
1-chloro-5-hexanone
CC(=O)CCCCCl
134.603
134.61

Chemical Identifiers

10226-30-9
134.603
CMDIDTNMHQUVPE-UHFFFAOYSA-N
82468
CC(=O)CCCCCl
C6H11ClO
MFCD00191625
1-chloro-5-hexanone
6-chlorohexan-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents