Learn More
6-Chloro-2-hexanone, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5346206
Description
6-Chloro-2-hexanone in the synthesis of the keto analogs began with 6-chloro-2-hexanone. Substituted 7-(oxoalkyl)theophyllines were synthesized by alkylation of the respective theophyllines with 6-chloro-2-hexanone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Chloro-2-hexanone in the synthesis of the keto analogs began with 6-chloro-2-hexanone. Substituted 7-(oxoalkyl)theophyllines were synthesized by alkylation of the respective theophyllines with 6-chloro-2-hexanone.
Solubility
Insoluble in water, soluble in chloroform (chloroform, carbon), n-heptane, ethanol, butanol, etc.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
6-Chloro-2-hexanone | |
1.02 | |
91°C (196°F) | |
1.4435 | |
5 g | |
Insoluble in water,soluble in chloroform (chloroform,carbon),n-heptane,ethanol,butanol,etc. | |
CC(=O)CCCCCl | |
134.603 | |
134.61 |
10226-30-9 | |
86°C (16 mmHg) | |
C6H11ClO | |
MFCD00191625 | |
6-chloro-2-hexanone, 1-chloro-5-hexanone, 2-hexanone, 6-chloro, 1-chlorohexan-5-one, 1-chlorohexane-5-one, 4-chlorobutyl methyl ketone, chlorohexanone, 1-chloro-hexan-5-one, 6-chloro-hexan-2-one, 6-chloranylhexan-2-one | |
CMDIDTNMHQUVPE-UHFFFAOYSA-N | |
6-chlorohexan-2-one | |
82468 | |
98% |
Chemical Identifiers
10226-30-9 | |
134.603 | |
CMDIDTNMHQUVPE-UHFFFAOYSA-N | |
82468 | |
CC(=O)CCCCCl |
C6H11ClO | |
MFCD00191625 | |
6-chloro-2-hexanone, 1-chloro-5-hexanone, 2-hexanone, 6-chloro, 1-chlorohexan-5-one, 1-chlorohexane-5-one, 4-chlorobutyl methyl ketone, chlorohexanone, 1-chloro-hexan-5-one, 6-chloro-hexan-2-one, 6-chloranylhexan-2-one | |
6-chlorohexan-2-one |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 233-546-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only