Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-2-fluoronicotinic acid, 95%

Chemical Identifiers

Chemical Name or Material 6-Chloro-2-fluoronicotinic acid
CAS 1211578-46-9
Formula Weight 175.54
MDL Number MFCD16610397
Synonym 6-Chloro-2-fluoropyridine-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6648403

 
alfa aesar™
H6648403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66484MD

 
alfa aesar™
H66484MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 175.54
Physical Form Solid
Assay 95%
MDL Number MFCD16610397
CAS 1211578-46-9
Synonym 6-Chloro-2-fluoropyridine-3-carboxylic acid
Chemical Name or Material 6-Chloro-2-fluoronicotinic acid
Quantity 1g