Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-2-fluoro-3-methylphenylacetic acid, 97%

$62.84 - $155.20

Chemical Identifiers

Chemical Name or Material 6-Chloro-2-fluoro-3-methylphenylacetic acid
CAS 261762-93-0
Molecular Formula C9H8ClFO2
Formula Weight 202.61
MDL Number MFCD01631375
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2663403

  SDS
alfa aesar™
H2663403
1g Each for $155.20

AAH26634MD

  SDS
alfa aesar™
H26634MD
250mg Each for $62.84
Description & Specifications

Specifications

Formula Weight 202.61
CAS 261762-93-0
MDL Number MFCD01631375
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C9H8ClFO2
Assay 97%
Quantity 1g
Chemical Name or Material 6-Chloro-2-fluoro-3-methylphenylacetic acid
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.