Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-2-fluoro-3-methylbenzyl bromide, 97%

Chemical Identifiers

Chemical Name or Material 6-Chloro-2-fluoro-3-methylbenzyl bromide
CAS 261762-88-3
Molecular Formula C8H7BrClF
Formula Weight 237.5
MDL Number MFCD01631373
UN Number UN1759
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2634006

 
alfa aesar™
H2634006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 237.5
Chemical Name or Material 6-Chloro-2-fluoro-3-methylbenzyl bromide
Assay 97%
Molecular Formula C8H7BrClF
MDL Number MFCD01631373
CAS 261762-88-3
UN Number UN1759
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
Boiling Point 103°C (3mmHg)
Melting Point 52° to 56°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.