Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-2-fluoro-3-methylbenzoic acid, 97%

$60.66 - $895.86

Chemical Identifiers

Chemical Name or Material 6-Chloro-2-fluoro-3-methylbenzoic acid
CAS 32890-90-7
Molecular Formula C8H6ClFO2
Formula Weight 188.59
MDL Number MFCD01631369
Beilstein 2259188
Synonym 6-Chloro-2-fluoro-m-toluic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2309503

 
alfa aesar™
B2309503
1g Each for $60.66

AAB2309514

 
alfa aesar™
B2309514
25g Each for $895.86
Description & Specifications

Specifications

Formula Weight 188.59
Chemical Name or Material 6-Chloro-2-fluoro-3-methylbenzoic acid
Assay 97%
Molecular Formula C8H6ClFO2
MDL Number MFCD01631369
CAS 32890-90-7
Beilstein 2259188
Synonym 6-Chloro-2-fluoro-m-toluic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 135° to 136°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.