Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-2-fluoro-3-methylaniline, 97%

$130.84 - $392.51

Chemical Identifiers

Chemical Name or Material 6-Chloro-2-fluoro-3-methylaniline
Molecular Formula C7H7ClFN
Formula Weight 159.59
MDL Number MFCD06660187
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2675703

 
alfa aesar™
H2675703
1g Each for $130.84

AAH2675706

 
alfa aesar™
H2675706
5g Each for $392.51
Description & Specifications

Specifications

Formula Weight 159.59
Chemical Name or Material 6-Chloro-2-fluoro-3-methylaniline
Assay 97%
Molecular Formula C7H7ClFN
MDL Number MFCD06660187
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.