Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-2-fluoro-3-methoxybenzylamine, 97%

Chemical Identifiers

Chemical Name or Material 6-Chloro-2-fluoro-3-methoxybenzylamine
Molecular Formula C8H9ClFNO
Formula Weight 189.62
MDL Number MFCD09832379
UN Number UN3259
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3154603

  SDS
alfa aesar™
H3154603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3154606

  SDS
alfa aesar™
H3154606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 189.62
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Assay 97%
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Quantity 1g
UN Number UN3259
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
Melting Point 55° to 57°C
Molecular Formula C8H9ClFNO
MDL Number MFCD09832379
Chemical Name or Material 6-Chloro-2-fluoro-3-methoxybenzylamine

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents, air, carbon dioxide. Store under inert gas.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.