missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-2-fluoro-3-methoxybenzamide, 97%

$47.09 - $47.09

Chemical Identifiers

CAS 886761-75-7
Molecular Formula C8H7ClFNO2
Molecular Weight (g/mol) 203.597
MDL Number MFCD06660354
InChI Key AZUOJQIOSGRMMS-UHFFFAOYSA-N
Synonym 6-chloro-2-fluoro-3-methoxybenzenecarboxamide
PubChem CID 17999360
IUPAC Name 6-chloro-2-fluoro-3-methoxybenzamide
SMILES COC1=C(C(=C(C=C1)Cl)C(=O)N)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3163803
View Documents
Alfa Aesar™
H3163803
1g
Each for $47.09
 
Description

Description

Specifications

Chemical Identifiers

886761-75-7
203.597
AZUOJQIOSGRMMS-UHFFFAOYSA-N
17999360
COC1=C(C(=C(C=C1)Cl)C(=O)N)F
C8H7ClFNO2
MFCD06660354
6-chloro-2-fluoro-3-methoxybenzenecarboxamide
6-chloro-2-fluoro-3-methoxybenzamide

Specifications

143°C to 145°C
886761-75-7
MFCD06660354
AZUOJQIOSGRMMS-UHFFFAOYSA-N
6-chloro-2-fluoro-3-methoxybenzamide
17999360
97%
1g
C8H7ClFNO2
6-chloro-2-fluoro-3-methoxybenzenecarboxamide
COC1=C(C(=C(C=C1)Cl)C(=O)N)F
203.597
203.6
6-Chloro-2-fluoro-3-methoxybenzamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Product Certifications