Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-2-fluoro-3-methoxybenzaldehyde, 97%

$135.31 - $760.41

Chemical Identifiers

Chemical Name or Material 6-Chloro-2-fluoro-3-methoxybenzaldehyde
CAS 112641-64-2
Molecular Formula C8H6ClFO2
Formula Weight 188.59
MDL Number MFCD04115946
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2644403

 
alfa aesar™
H2644403
1g Each for $135.31

AAH2644409

 
alfa aesar™
H2644409
10g Each for $760.41
Description & Specifications

Specifications

Formula Weight 188.59
Chemical Name or Material 6-Chloro-2-fluoro-3-methoxybenzaldehyde
Assay 97%
Molecular Formula C8H6ClFO2
MDL Number MFCD04115946
CAS 112641-64-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 95° to 100°C
Quantity 1g

Applications
Used as solvents. They are used in intermediate steps for the production of paints, plastics, synthetic resins, and dyes. They are also used in the manufacture of perfumes, solvents, and flavorings.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air and reducing agents. Store under an inert atmosphere.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.