Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-2-fluoro-3-methoxyaniline, 97%

Chemical Identifiers

Chemical Name or Material 6-Chloro-2-fluoro-3-methoxyaniline
CAS 1017777-77-3
Molecular Formula C7H7ClFNO
Formula Weight 175.59
MDL Number MFCD09832271
UN Number UN2811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3177906

  SDS
alfa aesar™
H3177906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 175.59
MDL Number MFCD09832271
Molecular Formula C7H7ClFNO
Chemical Name or Material 6-Chloro-2-fluoro-3-methoxyaniline
UN Number UN2811
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Quantity 5g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Health Hazard 2 GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 1 Danger
Assay 97%
CAS 1017777-77-3
Melting Point 40° to 43°C

Applications
6-Chloro-2-fluoro-3-methoxyaniline is used as a protected form of ammonia in a practical synthesis of azetidine

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.