missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Chloro-2,4-difluoroaniline, 97%, ACROS Organics™

Chemical Identifiers

CAS 36556-56-6
Molecular Formula C6H4ClF2N
Molecular Weight (g/mol) 163.55
MDL Number MFCD00142892
InChI Key DUPRZIYJXCCXQZ-UHFFFAOYSA-N
Synonym 2,4-difluoro-6-chloroaniline, 2-amino-3,5-difluorochlorobenzene, 2-chloro-4,6 difluoro aniline, 2-chloro-4,6-difluorophenylamine, 4,6-difluoro-2-chloroaniline, 6-chloro-2,4-difluoroaniline, acmc-1aj0r, benzenamine, 2-chloro-4,6-difluoro, ksc226e0f, pubchem2940
PubChem CID 2734083
IUPAC Name 2-chloro-4,6-difluoroaniline
SMILES C1=C(C=C(C(=C1F)N)Cl)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC350200010
SDS View Documents
Thermo Fisher Scientific
350200010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC350200050
SDS View Documents
Thermo Fisher Scientific
350200050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

36556-56-6
163.55
DUPRZIYJXCCXQZ-UHFFFAOYSA-N
2734083
C1=C(C=C(C(=C1F)N)Cl)F
C6H4ClF2N
MFCD00142892
2,4-difluoro-6-chloroaniline, 2-amino-3,5-difluorochlorobenzene, 2-chloro-4,6 difluoro aniline, 2-chloro-4,6-difluorophenylamine, 4,6-difluoro-2-chloroaniline, 6-chloro-2,4-difluoroaniline, acmc-1aj0r, benzenamine, 2-chloro-4,6-difluoro, ksc226e0f, pubchem2940
2-chloro-4,6-difluoroaniline

Specifications

Authentic
Gray-Green to White
1g
96.0
96% min. (GC)
MFCD00142892
DUPRZIYJXCCXQZ-UHFFFAOYSA-N
2-chloro-4,6-difluoroaniline
2734083
Low Melting Solid
6-Chloro-2, 4-difluoroaniline, 97%
Glass bottle
28°C to 33°C
36556-56-6
100.0
C6H4ClF2N
2,4-difluoro-6-chloroaniline, 2-amino-3,5-difluorochlorobenzene, 2-chloro-4,6 difluoro aniline, 2-chloro-4,6-difluorophenylamine, 4,6-difluoro-2-chloroaniline, 6-chloro-2,4-difluoroaniline, acmc-1aj0r, benzenamine, 2-chloro-4,6-difluoro, ksc226e0f, pubchem2940
C1=C(C=C(C(=C1F)N)Cl)F
163.55
163.55
97%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS