Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-2,3-dihydroimidazo[1,2-b]pyridazine, 98%

Chemical Identifiers

Chemical Name or Material 6-Chloro-2,3-dihydroimidazo[1,2-b]pyridazine
CAS 24439-00-7
Formula Weight 155.58
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6152703

 
alfa aesar™
H6152703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6152706

 
alfa aesar™
H6152706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 155.58
Chemical Name or Material 6-Chloro-2,3-dihydroimidazo[1,2-b]pyridazine
Physical Form Solid
Assay 98%
CAS 24439-00-7
Quantity 1g