missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-2,3-difluorobenzoyl chloride, 97%

Manufacturer:  Alfa Aesar™ H3387503

 View more versions of this product

Catalog No. AAH3387503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

6-Chloro-2,3-difluorobenzoyl chloride
1g
C7H2Cl2F2O
UN3265
C1=CC(=C(C(=C1F)F)C(=O)Cl)Cl
210.989
211
97%
Moisture Sensitive
1208078-25-4
MFCD12922578
CHLDUAMDKJNJKC-UHFFFAOYSA-N
6-chloro-2,3-difluorobenzoyl chloride
50998364
Liquid

Chemical Identifiers

1208078-25-4
210.989
CHLDUAMDKJNJKC-UHFFFAOYSA-N
6-chloro-2,3-difluorobenzoyl chloride
C7H2Cl2F2O
MFCD12922578
50998364
C1=CC(=C(C(=C1F)F)C(=O)Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

SDS Documents