missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-2,3-difluorobenzaldehyde, 97%

Manufacturer:  Alfa Aesar™ H3428706

 View more versions of this product

Catalog No. AAH3428706


 

Specifications

Specifications

6-Chloro-2,3-difluorobenzaldehyde
White through Yellow
797791-33-4
C7H3ClF2O
YVKZXQCYLHFHIR-UHFFFAOYSA-N
6-chloro-2,3-difluorobenzaldehyde
9812909
Solid
Air Sensitive
5g
97%
MFCD07368118
C1=CC(=C(C(=C1F)F)C=O)Cl
176.547
176.55

Chemical Identifiers

797791-33-4
176.547
YVKZXQCYLHFHIR-UHFFFAOYSA-N
C7H3ClF2O
MFCD07368118
9812909
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents