missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(6-Chloro-1H-benzotriazol-1-yloxy)tripyrrolidinophosphonium Hexafluorophosphate, TCI America™

Manufacturer:  TCI America C346125G

 View more versions of this product

Catalog No. C346125G



Specifications

Specifications

(6-Chloro-1H-benzotriazol-1-yloxy)tripyrrolidinophosphonium Hexafluorophosphate
173°C
893413-42-8
MFCD14560589
QJZCQEPNBRAYQL-UHFFFAOYSA-N
(6-chlorobenzotriazol-1-yl)oxy-tripyrrolidin-1-ylphosphanium;hexafluorophosphate
16039314
Crystal-Powder at 20°C
White-Yellow
25g
C18H27ClF6N6OP2
TPTDP
C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)ON4C5=C(C=CC(=C5)Cl)N=N4.F[P-](F)(F)(F)(F)F
554.843
554.84
≥98.0% (HPLC)

Chemical Identifiers

893413-42-8
554.843
QJZCQEPNBRAYQL-UHFFFAOYSA-N
16039314
C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)ON4C5=C(C=CC(=C5)Cl)N=N4.F[P-](F)(F)(F)(F)F
C18H27ClF6N6OP2
MFCD14560589
TPTDP
(6-chlorobenzotriazol-1-yl)oxy-tripyrrolidin-1-ylphosphanium;hexafluorophosphate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents