missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Chloro-1-hydroxy-1H-benzotriazole, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC448920050
Specifications
6-Chloro-1-hydroxy-1H-benzotriazole | |
190.0°C to 195.0°C | |
26198-19-6 | |
C6H4ClN3O | |
TZCYLJGNWDVJRA-UHFFFAOYSA-N | |
6-chloro-1-hydroxybenzotriazole | |
232711 | |
97% |
Glass bottle | |
5g | |
96% min. (HPLC) | |
1-hydroxy-6-chlorobenzotriazole, 1h-benzotriazole, 6-chloro-1-hydroxy, 6-chloro-1,2,3-benzotriazol-1-ol, 6-chloro-1-hydroxibenzotriazol, 6-chloro-1-hydroxy-1h-benzotriazole, 6-chloro-1-hydroxybenzotriazol, 6-chloro-1h-1,2,3-benzotriazol-1-ol, 6-chloro-1h-benzo d 1,2,3 triazol-1-ol, 6-chloro-1h-benzotriazol-1-ol, cl-hobt | |
C1=CC2=C(C=C1Cl)N(N=N2)O | |
169.57 | |
169.57 |
Chemical Identifiers
26198-19-6 | |
169.57 | |
1-hydroxy-6-chlorobenzotriazole, 1h-benzotriazole, 6-chloro-1-hydroxy, 6-chloro-1,2,3-benzotriazol-1-ol, 6-chloro-1-hydroxibenzotriazol, 6-chloro-1-hydroxy-1h-benzotriazole, 6-chloro-1-hydroxybenzotriazol, 6-chloro-1h-1,2,3-benzotriazol-1-ol, 6-chloro-1h-benzo d 1,2,3 triazol-1-ol, 6-chloro-1h-benzotriazol-1-ol, cl-hobt | |
6-chloro-1-hydroxybenzotriazole |
C6H4ClN3O | |
TZCYLJGNWDVJRA-UHFFFAOYSA-N | |
232711 | |
C1=CC2=C(C=C1Cl)N(N=N2)O |
Safety and Handling
GHS Signal Word: Warning