missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-1-hexyne, 98%

Manufacturer:  Alfa Aesar™ H53476AC

 View more versions of this product

Catalog No. AAH53476AC



Description

Description

Applications
6-Chloro-1-hexyne is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications

Specifications

6-Chloro-1-hexyne
37°C (99°F)
83°C (95mmHg)
10297-06-0
Cl(CH2)4C≡CH
UN1993
ZUKOCGMVJUXIJA-UHFFFAOYSA-N
6-chlorohex-1-yne
272001
98%
0.962
1.451
25mL
C6H9Cl
MFCD00013697
1-chloro-5-hexyne
C#CCCCCCl
116.588
116.59

Chemical Identifiers

10297-06-0
116.588
ZUKOCGMVJUXIJA-UHFFFAOYSA-N
272001
C#CCCCCCl
C6H9Cl
MFCD00013697
1-chloro-5-hexyne
6-chlorohex-1-yne
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents