Please login to your online account to display your discounted pricing

6-chloro-1-hexene 97%, ACROS Organics™

$213.20 - $213.20

Chemical Identifiers

Chemical Name or Material 6-Chloro-1-hexene
CAS 928-89-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H11Cl
Linear Formula CH2CH(CH2)4Cl
Formula Weight 118.61
MDL Number MFCD00039391
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC380920050

  SDS
thermo fisher scientific
AC380920050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC380920250

  SDS
thermo fisher scientific
AC380920250
Glass bottle 25g Each for $213.20
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 118.61
Quantity 5g
Color Clear colorless to light yellow
Density 0.9070g/mL
CAS Max % 100.0
CAS Min % 96.0
Packaging Glass bottle
Refractive Index 1.4360 to 1.4380 (20°C, 589nm)
Infrared Spectrum Authentic
Linear Formula CH2CH(CH2)4Cl
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Assay Percent Range 97%
MDL Number MFCD00039391
Health Hazard 1 GHS Signal Word: Warning
Molecular Formula C6H11Cl
Chemical Name or Material 6-Chloro-1-hexene
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 928-89-2
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.