Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-1,4-dihydro-2,3-quinoxalinedione, 97%

$39.62 - $455.04

Chemical Identifiers

Chemical Name or Material 6-Chloro-1,4-dihydro-2,3-quinoxalinedione
CAS 169-14-2
Molecular Formula C8H5ClN2O2
Formula Weight 196.59
MDL Number MFCD00140720
Beilstein 168142
Synonym 6-Chloro-2,3-dihydroxyquinoxaline; 6-Chloro-2,3-dioxo-1,2,3,4-tetrahydroquinoxaline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1403803

  SDS
alfa aesar™
L1403803
1g Each for $39.62

AAL1403806

  SDS
alfa aesar™
L1403806
5g Each for $117.38

AAL1403814

  SDS
alfa aesar™
L1403814
25g Each for $455.04
Description & Specifications

Specifications

Formula Weight 196.59
MDL Number MFCD00140720
Health Hazard 1 Warning
CAS 169-14-2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 6-Chloro-1,4-dihydro-2,3-quinoxalinedione
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
TSCA TSCA
Synonym 6-Chloro-2,3-dihydroxyquinoxaline; 6-Chloro-2,3-dioxo-1,2,3,4-tetrahydroquinoxaline
Molecular Formula C8H5ClN2O2
Assay 97%
Quantity 1g
Melting Point >300°C
Beilstein 168142
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.