missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Bromothiochroman-4-one, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3196806

 View more versions of this product

Catalog No. AAH3196806



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in cool dry place. Keep away from oxidizing agents.
Specifications

Specifications

6-Bromothiochroman-4-one
66°C to 69°C
MFCD05664869
6-bromothiochroman-4-one, 6-bromo-2,3-dihydro-4h-1-benzothiopyran-4-one, 6-bromo-3,4-dihydro-2h-1-benzothiopyran-4-one, 6-bromo-2,3-dihydro-1-benzothiopyran-4-one, 6-bromo-3,4-dihydro-2h-1-benzothiin-4-one, 6-bromo-thiochroman-4-one, 6-bromo-2h-1-benzothiopyran-4 3h-one, 6-bromo-2,3-dihydro-4h-1-benzothiopyran4-one, 2,3-dihydro-6-bromo-4h-1-benzothiopyran-4-one, 4h-1-benzothiopyran-4-one, 6-bromo-2,3-dihydro
C1CSC2=C(C1=O)C=C(C=C2)Br
243.118
243.12
13735-13-2
C9H7BrOS
5 g
SBNPYDJTVGRDAA-UHFFFAOYSA-N
6-bromo-2,3-dihydrothiochromen-4-one
13953736
95%

Chemical Identifiers

13735-13-2
243.118
SBNPYDJTVGRDAA-UHFFFAOYSA-N
13953736
C1CSC2=C(C1=O)C=C(C=C2)Br
C9H7BrOS
MFCD05664869
6-bromothiochroman-4-one, 6-bromo-2,3-dihydro-4h-1-benzothiopyran-4-one, 6-bromo-3,4-dihydro-2h-1-benzothiopyran-4-one, 6-bromo-2,3-dihydro-1-benzothiopyran-4-one, 6-bromo-3,4-dihydro-2h-1-benzothiin-4-one, 6-bromo-thiochroman-4-one, 6-bromo-2h-1-benzothiopyran-4 3h-one, 6-bromo-2,3-dihydro-4h-1-benzothiopyran4-one, 2,3-dihydro-6-bromo-4h-1-benzothiopyran-4-one, 4h-1-benzothiopyran-4-one, 6-bromo-2,3-dihydro
6-bromo-2,3-dihydrothiochromen-4-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only