missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromoquinoline, 97%

Manufacturer:  Alfa Aesar™ H5511106

 View more versions of this product

Catalog No. AAH5511106



Description

Description

Applications
6-Bromoquinoline is used as fine chemical and pharmaceutical intermediate, used as the coupling reagent.

Solubility
Soluble in acetone, acetonitrile, dichloromethane, ethyl acetate and THF.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

6-Bromoquinoline
>110°C (230°F)
24°C
5332-25-2
MFCD00024023
Soluble in acetone,acetonitrile,dichloromethane,ethyl acetate and THF.
C1=CC2=C(C=CC(=C2)Br)N=C1
208.058
208.06
1.538
1.663
5g
C9H6BrN
6-br-quinoline
IFIHYLCUKYCKRH-UHFFFAOYSA-N
6-bromoquinoline
79243
97%

Chemical Identifiers

5332-25-2
208.058
IFIHYLCUKYCKRH-UHFFFAOYSA-N
79243
C1=CC2=C(C=CC(=C2)Br)N=C1
C9H6BrN
MFCD00024023
6-br-quinoline
6-bromoquinoline
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 226-238-7

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents