Please login to your online account to display your discounted pricing

6-Bromoquinoline, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material 6-Bromoquinoline
CAS 5332-25-2
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H6BrN
Formula Weight 208.06
MDL Number MFCD00024023
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC04510CB

 
fisher scientific company
CC04510CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 208.06
Quantity 250mg
Assay 95%
CAS Max % 100.0
Chemical Name or Material 6-Bromoquinoline
Molecular Formula C9H6BrN
CAS 5332-25-2
Assay Percent Range 95%
MDL Number MFCD00024023
CAS Min % 94.5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.