missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromoquinazoline-2,4(1H,3H)-dione, 97%

Manufacturer:  Alfa Aesar™ H63547MD

 View more versions of this product

Catalog No. AAH63547MD



Specifications

Specifications

6-Bromoquinazoline-2,4(1H,3H)-dione
250mg
C8H5BrN2O2
6-bromoquinazoline-2,4 1h,3h-dione, 6-bromo-2,4 1h,3h-quinazolinedione, 6-bromoquinazoline-2,4-dione, 6-bromoquinazoline-2,4-diol, 2,4 1h,3h-quinazolinedione, 6-bromo, 6-bromo-1,3-dihydroquinazoline-2,4-dione, 6-bromo-1,2,3,4-tetrahydroquinazoline-2,4-dione, bromoquinazolinedione, pubchem20961, acmc-209qre
C1=CC2=C(C=C1Br)C(=O)NC(=O)N2
241.044
241.04
97%
White
88145-89-5
MFCD00462868
JZDVFUAHGLJVQG-UHFFFAOYSA-N
6-bromo-1H-quinazoline-2,4-dione
617686
Solid

Chemical Identifiers

88145-89-5
241.044
JZDVFUAHGLJVQG-UHFFFAOYSA-N
617686
C1=CC2=C(C=C1Br)C(=O)NC(=O)N2
C8H5BrN2O2
MFCD00462868
6-bromoquinazoline-2,4 1h,3h-dione, 6-bromo-2,4 1h,3h-quinazolinedione, 6-bromoquinazoline-2,4-dione, 6-bromoquinazoline-2,4-diol, 2,4 1h,3h-quinazolinedione, 6-bromo, 6-bromo-1,3-dihydroquinazoline-2,4-dione, 6-bromo-1,2,3,4-tetrahydroquinazoline-2,4-dione, bromoquinazolinedione, pubchem20961, acmc-209qre
6-bromo-1H-quinazoline-2,4-dione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents