missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromopyridine-2-carboxaldehyde, 97%

$52.66 - $644.74

Chemical Identifiers

CAS 34160-40-2
Molecular Formula C6H4BrNO
Formula Weight 186.01
MDL Number MFCD02683546
Beilstein 1524300
Chemical Name or Material 6-Bromopyridine-2-carboxaldehyde
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1951803 SDS Alfa Aesar™
L1951803
1g Each for $52.66
AAL1951806 SDS Alfa Aesar™
L1951806
5g Each for $160.07
AAL1951814 SDS Alfa Aesar™
L1951814
25g Each for $644.74
Specifications

Description

Applications
6-Bromopyridine-2-carboxaldehyde is used in the study of a rhodium-catalyzed, reductive aldol coupling with divinyl ketones leading to syn ß-hydroxyenones. It is also used as a building block for Tris[(pyridyl)methyl]amine ligands.

Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol. Insoluble in water.

Notes
Store under inert gas. It is sensitive to air. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

34160-40-2
186.01
1524300
C6H4BrNO
MFCD02683546
6-Bromopyridine-2-carboxaldehyde

Specifications

34160-40-2
186.01
79° to 83°C
1g
97%
C6H4BrNO
MFCD02683546
1524300
6-Bromopyridine-2-carboxaldehyde

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning