missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromooxindole, 97%

Manufacturer:  Alfa Aesar™ H5601206

 View more versions of this product

Catalog No. AAH5601206



Description

Description

Applications
6-Bromooxindole derivative is used in the preparation of p38alpha inhibitors as potential anti- inflammatories. It is used as an intermediate in pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

6-Bromooxindole
99365-40-9
MFCD02179605
Slightly soluble in water.
C1C2=C(C=C(C=C2)Br)NC1=O
212.046
212.05
5g
C8H6BrNO
2h-indol-2-one, 6-bromo-1,3-dihydro, 6-bromo-1,3-dihydro-2h-indol-2-one, 6-bromo-1,3-dihydro-indol-2-one, 6-bromo-2,3-dihydro-1h-indol-2-one, 6-bromo-2-indolinone, 6-bromo-2-oxindole, 6-bromo-2-oxyindole, 6-bromoindolin-2-one, 6-bromooxindol, 6-bromooxindole
JARRYVQFBQVOBE-UHFFFAOYSA-N
6-bromo-1,3-dihydroindol-2-one
2773289
97%

Chemical Identifiers

99365-40-9
212.046
JARRYVQFBQVOBE-UHFFFAOYSA-N
2773289
C1C2=C(C=C(C=C2)Br)NC1=O
C8H6BrNO
MFCD02179605
2h-indol-2-one, 6-bromo-1,3-dihydro, 6-bromo-1,3-dihydro-2h-indol-2-one, 6-bromo-1,3-dihydro-indol-2-one, 6-bromo-2,3-dihydro-1h-indol-2-one, 6-bromo-2-indolinone, 6-bromo-2-oxindole, 6-bromo-2-oxyindole, 6-bromoindolin-2-one, 6-bromooxindol, 6-bromooxindole
6-bromo-1,3-dihydroindol-2-one
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents