missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Bromonicotinic acid, 97%

$190.86 - $626.49

Chemical Identifiers

CAS 6311-35-9
Molecular Formula C6H4BrNO2
Formula Weight 202.01
MDL Number MFCD01927100
Synonym 6-Bromopyridine-3-carboxylic acid
Chemical Name or Material 6-Bromonicotinic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2792606 SDS Alfa Aesar™
H2792606
5g Each for $190.86
AAH2792614 SDS Alfa Aesar™
H2792614
25g Each for $626.49
Specifications Description & Specifications

Chemical Identifiers

6311-35-9
202.01
6-Bromopyridine-3-carboxylic acid
C6H4BrNO2
MFCD01927100
6-Bromonicotinic acid

Specifications

6311-35-9
5g
202.01
6-Bromopyridine-3-carboxylic acid
97%
179° to 188°C
C6H4BrNO2
MFCD01927100
6-Bromonicotinic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning