Please login to your online account to display your discounted pricing

6-(Bromomethyl)-4-chloro-2-(trifluoromethyl)quinoline, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material 6-(Bromomethyl)-4-chloro-2-(trifluoromethyl)quinoline
CAS 123637-51-4
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C11H6BrClF3N
Formula Weight 324.53
MDL Number MFCD00153079
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM07330EA

  SDS
fisher scientific company
KM07330EA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 324.53
Quantity 10g
Assay 95%
CAS Min % 95.0
Assay Percent Range 95%
Chemical Name or Material 6-(Bromomethyl)-4-chloro-2-(trifluoromethyl)quinoline
Molecular Formula C11H6BrClF3N
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
MDL Number MFCD00153079
CAS 123637-51-4
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.