missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-(Bromomethyl)-3,4-dihydro-2H-1,5-benzodioxepine, 97%, Maybridge™

Manufacturer:  Fisher Scientific Company CC00408DA

 View more versions of this product

Catalog No. CC00408DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

6-(Bromomethyl)-3, 4-dihydro-2H-1, 5-benzodioxepine
1g
96.5
97%
MFCD05664399
QDRNTKFXSFDJCH-UHFFFAOYSA-N
6-(bromomethyl)-3,4-dihydro-2H-1,5-benzodioxepine
2794992
97%
Amber Glass Bottle
499770-96-6
100.0
C10H11BrO2
2h-1,5-benzodioxepin, 6-bromomethyl-3,4-dihydro, 6-bromomethyl-2h,3h,4h-benzo b 1,4-dioxepin, 6-bromomethyl-3,4-dihydro-1,5-benzodioxepin, 6-bromomethyl-3,4-dihydro-2h-1,5-benzodioxepine
C1COC2=CC=CC(=C2OC1)CBr
243.1
243.1

Chemical Identifiers

499770-96-6
243.1
QDRNTKFXSFDJCH-UHFFFAOYSA-N
2794992
C1COC2=CC=CC(=C2OC1)CBr
C10H11BrO2
MFCD05664399
2h-1,5-benzodioxepin, 6-bromomethyl-3,4-dihydro, 6-bromomethyl-2h,3h,4h-benzo b 1,4-dioxepin, 6-bromomethyl-3,4-dihydro-1,5-benzodioxepin, 6-bromomethyl-3,4-dihydro-2h-1,5-benzodioxepine
6-(bromomethyl)-3,4-dihydro-2H-1,5-benzodioxepine
Safety and Handling

Safety and Handling

GHS P Statement Causes severe skin burns and eye damage.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair):

Danger

SDS Documents