Please login to your online account to display your discounted pricing

6-(Bromomethyl)-3,4-dihydro-2H-1,5-benzodioxepine, 97%, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  CC00408DE

 View more versions of this product

Catalog No. CC00408DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 243.1
Quantity 5g
Assay 97%
CAS Max % 100.0
Health Hazard 1 Danger
CAS 499770-96-6
Molecular Formula C10H11BrO2
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair):
CAS Min % 96.5
MDL Number MFCD05664399
Assay Percent Range 97%
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage.
Chemical Name or Material 6-(Bromomethyl)-3, 4-dihydro-2H-1, 5-benzodioxepine

Specifications

Chemical Name or Material 6-(Bromomethyl)-3, 4-dihydro-2H-1, 5-benzodioxepine
CAS 499770-96-6
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H11BrO2
Formula Weight 243.1
MDL Number MFCD05664399
Accessories

Danger

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair):

GHS P Statement Causes severe skin burns and eye damage.