missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-(Bromomethyl)-2-methylquinoline, ≥97%, Maybridge

Chemical Identifiers

CAS 141848-60-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H10BrN
Formula Weight 236.11
MDL Number MFCD06658974
Chemical Name or Material 6-(Bromomethyl)-2-methylquinoline
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC28408CB SDS Fisher Scientific Company
CC28408CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC28408DA SDS Fisher Scientific Company
CC28408DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

141848-60-4
100.0
C11H10BrN
MFCD06658974
97%
96.5
97%
236.11
6-(Bromomethyl)-2-methylquinoline

Specifications

141848-60-4
100.0
250mg
C11H10BrN
MFCD06658974
97%
96.5
Amber Glass Bottle
97%
236.11
6-(Bromomethyl)-2-methylquinoline

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger