missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Bromoimidazo[1,2-a]pyrimidine, TCI America™

$123.75 - $426.27

Chemical Identifiers

CAS 865156-68-9
Molecular Formula C6H4BrN3
Molecular Weight (g/mol) 198.023
MDL Number MFCD09261434
InChI Key BQMWMOQCMFLRQQ-UHFFFAOYSA-N
Synonym 6-bromoimidazo 1,2-a pyrimidine, 6-bromo-imidazo 1,2-a pyrimidine, imidazo 1,2-a pyrimidine, 6-bromo, 6-bromo-4-hydroimidazo 1,2-a pyrimidine, pubchem20931, acmc-209qa4, 6-bromo-imidazo 1,2-a pyr..., 6-bromanylimidazo 1,2-a pyrimidine, imidazo 1,2-a pyrimidine,6-bromo, 6-bromoimidazo 1,2-a pyrimidine
PubChem CID 26967622
IUPAC Name 6-bromoimidazo[1,2-a]pyrimidine
SMILES C1=CN2C=C(C=NC2=N1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B55991G View Documents TCI America
B55991G
1g Each for $123.75
 
B55995G View Documents TCI America
B55995G
5g Each for $426.27
 
Specifications

Chemical Identifiers

865156-68-9
198.023
BQMWMOQCMFLRQQ-UHFFFAOYSA-N
26967622
C1=CN2C=C(C=NC2=N1)Br
C6H4BrN3
MFCD09261434
6-bromoimidazo 1,2-a pyrimidine, 6-bromo-imidazo 1,2-a pyrimidine, imidazo 1,2-a pyrimidine, 6-bromo, 6-bromo-4-hydroimidazo 1,2-a pyrimidine, pubchem20931, acmc-209qa4, 6-bromo-imidazo 1,2-a pyr..., 6-bromanylimidazo 1,2-a pyrimidine, imidazo 1,2-a pyrimidine,6-bromo, 6-bromoimidazo 1,2-a pyrimidine
6-bromoimidazo[1,2-a]pyrimidine

Specifications

White-Yellow
1g
C6H4BrN3
6-bromoimidazo 1,2-a pyrimidine, 6-bromo-imidazo 1,2-a pyrimidine, imidazo 1,2-a pyrimidine, 6-bromo, 6-bromo-4-hydroimidazo 1,2-a pyrimidine, pubchem20931, acmc-209qa4, 6-bromo-imidazo 1,2-a pyr..., 6-bromanylimidazo 1,2-a pyrimidine, imidazo 1,2-a pyrimidine,6-bromo, 6-bromoimidazo 1,2-a pyrimidine
C1=CN2C=C(C=NC2=N1)Br
198.023
198.02
≥98.0% (GC,T)
218°C
865156-68-9
MFCD09261434
BQMWMOQCMFLRQQ-UHFFFAOYSA-N
6-bromoimidazo[1,2-a]pyrimidine
26967622
Crystal-Powder at 20°C
6-Bromoimidazo[1,2-a]pyrimidine
Safety and Handling

Safety and Handling

TSCA : No