missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (6-Bromohexyloxy)-tert-butyldimethylsilane, 99%

Manufacturer:  Alfa Aesar™ H55581AA

 View more versions of this product

Catalog No. AAH55581AA



Specifications

Specifications

(6-Bromohexyloxy)-tert-butyldimethylsilane
C12H27BrOSi
1-t-butyldimethylsilyloxy-6-bromohexane
CC(C)(C)[Si](C)(C)OCCCCCCBr
295.336
295.34
99%
1.457
Colorless
110°C (230°F)
129368-70-3
MFCD01863708
PBKXRKYUUXKNSL-UHFFFAOYSA-N
6-bromohexoxy-tert-butyl-dimethylsilane
4260352
Liquid
1.053 g/mL
276°C
5mL

Chemical Identifiers

129368-70-3
295.336
PBKXRKYUUXKNSL-UHFFFAOYSA-N
4260352
CC(C)(C)[Si](C)(C)OCCCCCCBr
C12H27BrOSi
MFCD01863708
1-t-butyldimethylsilyloxy-6-bromohexane
6-bromohexoxy-tert-butyl-dimethylsilane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents