Learn More
6-Bromohexanoyl chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1106306
Description
6-Bromohexanoyl chloride is used as intermediates, organic synthesis, chemical research, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Bromohexanoyl chloride is used as intermediates, organic synthesis, chemical research, agrochemicals and dyestuff.
Solubility
Reacts with water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed. Moisture Sensitive
Specifications
6-Bromohexanoyl chloride | |
1.397 | |
>110°C (230°F) | |
C6H10BrClO | |
Br(CH2)5COCl | |
5 g | |
1749739 | |
hexanoyl chloride, 6-bromo, 6-bromohexanoic acid chloride, 6-bromohexanoylchloride, 6-bromocaproyl chloride, bromohexanoyl chloride, zlchem 435, acmc-209fyy, 6-bromo-hexanoylchloride, 6-bromo-hexanoyl chloride, 6-bromanylhexanoyl chloride | |
HBPVGJGBRWIVSX-UHFFFAOYSA-N | |
6-bromohexanoyl chloride | |
89848 | |
97% |
22809-37-6 | |
119°C to 120°C (9 mmHg) | |
Acrid | |
1.4875 | |
MFCD00000761 | |
UN3265 | |
Moisture sensitive | |
Reacts with water. | |
C(CCC(=O)Cl)CCBr | |
213.499 | |
213.5 |
Chemical Identifiers
22809-37-6 | |
213.499 | |
HBPVGJGBRWIVSX-UHFFFAOYSA-N | |
89848 | |
C(CCC(=O)Cl)CCBr |
C6H10BrClO | |
MFCD00000761 | |
hexanoyl chloride, 6-bromo, 6-bromohexanoic acid chloride, 6-bromohexanoylchloride, 6-bromocaproyl chloride, bromohexanoyl chloride, zlchem 435, acmc-209fyy, 6-bromo-hexanoylchloride, 6-bromo-hexanoyl chloride, 6-bromanylhexanoyl chloride | |
6-bromohexanoyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 245-236-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only