missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Bromohexanoic acid, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC165675000

 View more versions of this product

Catalog No. AC165675000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

6-Bromohexanoic acid
Authentic
165.0°C to 170.0°C (20.0mmHg)
32.0°C to 35.0°C
4224-70-8
C6H11BrO2
6-bromocaproic acid, hexanoic acid, 6-bromo, 6-bromo-hexanoic acid, 6-bromo-n-caproic acid, 6-bromo hexanoic acid, epsilon-bromo caproic acid, 6-bromo hexanoicacid, 6bromohexanoic acid, 6-bromo-hexanicacid, 6-bromo-caproic acid
C(CCC(=O)O)CCBr
195.056
CHEBI:60700
Low Melting Crystals
67°C
Glass bottle
Orange to White
500g
98%
Br(CH2)5CO2H
NVRVNSHHLPQGCU-UHFFFAOYSA-N
6-bromohexanoic acid
20210
195.06
98%

Chemical Identifiers

4224-70-8
195.056
6-bromocaproic acid, hexanoic acid, 6-bromo, 6-bromo-hexanoic acid, 6-bromo-n-caproic acid, 6-bromo hexanoic acid, epsilon-bromo caproic acid, 6-bromo hexanoicacid, 6bromohexanoic acid, 6-bromo-hexanicacid, 6-bromo-caproic acid
CHEBI:60700
C(CCC(=O)O)CCBr
C6H11BrO2
NVRVNSHHLPQGCU-UHFFFAOYSA-N
20210
6-bromohexanoic acid
SDS Documents