Learn More
6-Bromohexanenitrile, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1040009
Description
2,6-Lutidine is used as a solvent in organic synthesis and as a sterically hindered mild base. It is also used as a vulcanization accelerator for dyes, resins and rubber. It also acts as a food additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
6-Bromohexanenitrile | |
1.328 | |
>110°C (230°F) | |
1.4775 | |
10 g | |
1744172 | |
PHOSWLARCIBBJZ-UHFFFAOYSA-N | |
6-bromohexanenitrile | |
81093 | |
97% |
6621-59-6 | |
134°C to 135°C (15 mmHg) | |
C6H10BrN | |
MFCD00013834 | |
UN3276 | |
6-bromocapronitrile, 6-bromohexanonitrile, hexanenitrile, 6-bromo, 5-bromopentyl cyanide, 6-bromo-hexanenitrile, 6-bromanylhexanenitrile, pubchem12505, acmc-1b5au, 6-bromohexanenitrile, phoswlarcibbjz-uhfffaoysa | |
BrCCCCCC#N | |
176.06 | |
176.06 |
Chemical Identifiers
6621-59-6 | |
176.06 | |
PHOSWLARCIBBJZ-UHFFFAOYSA-N | |
81093 | |
BrCCCCCC#N |
C6H10BrN | |
MFCD00013834 | |
6-bromocapronitrile, 6-bromohexanonitrile, hexanenitrile, 6-bromo, 5-bromopentyl cyanide, 6-bromo-hexanenitrile, 6-bromanylhexanenitrile, pubchem12505, acmc-1b5au, 6-bromohexanenitrile, phoswlarcibbjz-uhfffaoysa | |
6-bromohexanenitrile |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber : 229-570-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only